missing translation for 'onlineSavingsMsg'
Learn More

ethyle1-(3-aminobenzyl)piperidine-4-carboxylate, 97%, Maybridge

Brand:  Maybridge SPB08422EA

Code : 31

Additional Details : CAS Number : 306937-22-4 Weight : 0.06000kg Transport : UN number : 3259 Chem class : 8 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 10482644

  • £193.00 / 10g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

ethyle1-(3-aminobenzyl)piperidine-4-carboxylate
97.0
97%
262.35
306937-22-4
97%
C15H22N2O2
MFCD02180894

Specifications

ethyle1-(3-aminobenzyl)piperidine-4-carboxylate
306937-22-4
97%
C15H22N2O2
MFCD02180894
10g
97.0
97%
262.35
Videos
SDS