ethyle(S)-3-hydroxybutyrate, 99%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 335260050

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 56816-01-4 Weight: 0.02700kg


Packaging Glass bottle
Quantity 5g

Product Code. 10177803

Quantity Price
1 £ 187.0 / 5g
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethyl (S)-3-hydroxybutyrate
Assay 99%
Assay Percent Range 99%
Molecular Formula C6H12O3
Linear Formula CH3CH(OH)CH2COOCH2CH3
Formula Weight 132.16
MDL Number MFCD00066206

Specification

Physical Form Liquid
Formula Weight 132.16
Quantity 5g
Color Clear colorless
Assay 99%
Specific Rotation Condition + 43.00 (20.00°C c=1,chloroform)
Linear Formula CH3CH(OH)CH2COOCH2CH3
Specific Rotation +43° to +47° (20°C, 589nm) (c=1, CHCl3)
Infrared Spectrum Authentic
CAS 56816-01-4
MDL Number MFCD00066206
Assay Percent Range 99%
Refractive Index 1.4195 to 1.4215 (20°C, 589nm)
Packaging Glass bottle
Molecular Formula C6H12O3
Density 1.0100g/mL
Chemical Name or Material Ethyl (S)-3-hydroxybutyrate
Documentation

Technical documents