missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-3-hydroxybutyrate, 99%, Acros Organics

£55.20 - £334.00

Chemical Identifiers

CAS 56816-01-4
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00066206
InChI Key OMSUIQOIVADKIM-YFKPBYRVSA-N
Synonym s-ethyl 3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, ethyl 3s-3-hydroxybutanoate, s-+-3-hydroxybutyric acid ethyl ester, ethyl s-+-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, ethyl s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl s---3-hydroxybutyrate
PubChem CID 6950306
IUPAC Name ethyl (3S)-3-hydroxybutanoate
SMILES CCOC(=O)CC(C)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11711151 View Documents Thermo Scientific
10011701
1g Glass bottle £334.00
Each View alternative products
Save Up to 
»
Estimated Shipment
16-07-2020
Add to basket
 
10011701 SDS View Documents Acros Organics
335260010
1g Glass bottle £55.20
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 17-06-2020
Please to purchase this item. Need a web account? Register with us today!
10177803 SDS View Documents Acros Organics
335260050
5g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Specifications

Chemical Identifiers

56816-01-4
132.16
OMSUIQOIVADKIM-YFKPBYRVSA-N
6950306
CCOC(=O)CC(C)O
C6H12O3
MFCD00066206
s-ethyl 3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, ethyl 3s-3-hydroxybutanoate, s-+-3-hydroxybutyric acid ethyl ester, ethyl s-+-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, ethyl s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl s---3-hydroxybutyrate
ethyl (3S)-3-hydroxybutanoate

Specifications

Ethyl (S)-3-hydroxybutyrate
64°C
Glass bottle
1.01
+ 43
Colorless
56816-01-4
C6H12O3
MFCD00066206
OMSUIQOIVADKIM-YFKPBYRVSA-N
ethyl (3S)-3-hydroxybutanoate
6950306
Liquid
1.0100g/mL
Authentic
1.4200 to 1.422
+ 43.00 (20.00°C c=1,chloroform)
180°C to 182°C
1g
98.5% min. (GC)
CH3CH(OH)CH2COOCH2CH3
s-ethyl 3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, ethyl 3s-3-hydroxybutanoate, s-+-3-hydroxybutyric acid ethyl ester, ethyl s-+-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, ethyl s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl s---3-hydroxybutyrate
CCOC(=O)CC(C)O
132.16
132.16
99%