missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 10mm Rectangular Cell Holder for 15mm z-height/BP
Brand: Thermo Scientific™ 10011701
Code : NEW
Additional Details : Weight : 0.01000kg
Specifications
+ 43 | |
1.4200 to 1.422 | |
Authentic | |
1g | |
Colorless | |
Glass bottle | |
56816-01-4 | |
C6H12O3 | |
MFCD00066206 | |
OMSUIQOIVADKIM-YFKPBYRVSA-N | |
ethyl (3S)-3-hydroxybutanoate | |
6950306 | |
Liquid |
1.0100g/mL | |
+ 43.00 (20.00°C c=1,chloroform) | |
64°C | |
1.01 | |
180°C to 182°C | |
Ethyl (S)-3-hydroxybutyrate | |
98.5% min. (GC) | |
CH3CH(OH)CH2COOCH2CH3 | |
s-ethyl 3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, ethyl 3s-3-hydroxybutanoate, s-+-3-hydroxybutyric acid ethyl ester, ethyl s-+-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, ethyl s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl s---3-hydroxybutyrate | |
CCOC(=O)CC(C)O | |
132.16 | |
132.16 | |
99% |