missing translation for 'onlineSavingsMsg'
Learn More

Ethyl alpha-bromophenylacetate, 97%, Thermo Scientific Chemicals

Catalog Number 11935001
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2882-19-1
C10H11BrO2
243.10
MFCD00013536
BKTKLDMYHTUESO-UHFFFAOYNA-N
ethyl alpha-bromophenylacetate, ethyl bromophenylacetate, ethyl bromo phenyl acetate, ethyl-alpha-bromophenyl acetate, ethyl-.alpha.-bromophenyl acetate, benzeneacetic acid, .alpha.-bromo-, ethyl ester, bromo-phenyl-acetic acid ethyl ester, benzeneacetic acid, alpha-bromo-, ethyl ester, eyr&vo2, ethylbromo phenyl acetate
97780
ethyl 2-bromo-2-phenylacetate
CCOC(=O)C(Br)C1=CC=CC=C1
This item is not returnable. View return policy
2882-19-1
C10H11BrO2
243.10
MFCD00013536
BKTKLDMYHTUESO-UHFFFAOYNA-N
ethyl alpha-bromophenylacetate, ethyl bromophenylacetate, ethyl bromo phenyl acetate, ethyl-alpha-bromophenyl acetate, ethyl-.alpha.-bromophenyl acetate, benzeneacetic acid, .alpha.-bromo-, ethyl ester, bromo-phenyl-acetic acid ethyl ester, benzeneacetic acid, alpha-bromo-, ethyl ester, eyr&vo2, ethylbromo phenyl acetate
97780
ethyl 2-bromo-2-phenylacetate
CCOC(=O)C(Br)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.3890g/mL
Boiling Point 89°C to 92°C (1.2 hPa)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5Ch(Br)COOCH2CH3
Refractive Index 1.5385 to 1.5405 (20°C, 589nm)
Quantity 5 g
Specific Gravity 1.389
Formula Weight 243.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl α-bromophenylacetate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl alpha-bromophenylacetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'