missing translation for 'onlineSavingsMsg'
Learn More

Ethylbenzene, 99.8%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4229210
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
100-41-4
C8H10
106.17
YNQLUTRBYVCPMQ-UHFFFAOYSA-N
phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
7500
CHEBI:16101
ethylbenzene
CCC1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
100-41-4
C8H10
106.17
YNQLUTRBYVCPMQ-UHFFFAOYSA-N
phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
7500
CHEBI:16101
ethylbenzene
CCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -95.0°C
Density 0.8600g/mL
Boiling Point 136.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 99.75% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula C6H5C2H5
Refractive Index 1.4940 to 1.4970
Quantity 100 mL
Residue after Evaporation 0.0005 % max.
Merck Index 15,382
Specific Gravity 0.86
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol,benzene,carbon tetrachloride,,ether and sulfur dioxide,slightly soluble in chloroform,insoluble in ammonia
Viscosity 0.63 mPa.s (20°C)
Formula Weight 106.17
Percent Purity 99.8%
Physical Form Liquid
Chemical Name or Material Ethylbenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethylbenzene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H304-May be fatal if swallowed and enters airways.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'