missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylbenzene, 99.8%, pure, Thermo Scientific Chemicals
27.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -95.0°C |
Density | 0.8600g/mL |
Boiling Point | 136.0°C |
Flash Point | 22°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99.7% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C8H10 |
Linear Formula | C6H5C2H5 |
Refractive Index | 1.4940 to 1.4970 |
Show More |
Safety and Handling
Product Identifier
- Ethylbenzene
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H225-Highly flammable liquid and vapour.
- H332-Harmful if inhaled.
- H304-May be fatal if swallowed and enters airways.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'