missing translation for 'onlineSavingsMsg'
Learn More

Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-3212702
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
563-43-9, 142-82-5
C2H5AlCl2
126.94
MFCD00000457
UAIZDWNSWGTKFZ-UHFFFAOYSA-L
ethylaluminum dichloride, aluminum, dichloroethyl, dichloroethylaluminum, ethyldichloroaluminum, dichloromonoethylaluminum, ethylaluminium dichloride, dichloro ethyl alumane, ethyl aluminum dichloride, hsdb 317, dichloro ethyl aluminum
dichloro(ethyl)alumane
CC[Al](Cl)Cl
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
563-43-9, 142-82-5
C2H5AlCl2
126.94
MFCD00000457
UAIZDWNSWGTKFZ-UHFFFAOYSA-L
ethylaluminum dichloride, aluminum, dichloroethyl, dichloroethylaluminum, ethyldichloroaluminum, dichloromonoethylaluminum, ethylaluminium dichloride, dichloro ethyl alumane, ethyl aluminum dichloride, hsdb 317, dichloro ethyl aluminum
dichloro(ethyl)alumane
CC[Al](Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.7400g/mL
Packaging AcroSeal™ Glass bottle
Linear Formula C2H5AlCl2
Quantity 100 mL
Fieser 06,251; 07,146; 10,177
Merck Index 15, 3816
Specific Gravity 0.74
Solubility Information Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons
Formula Weight 126.95
Percent Purity 14.5 to 15.5%
Physical Form Liquid
Chemical Name or Material Ethylaluminium dichloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'