missing translation for 'onlineSavingsMsg'
Learn More

Ethyl vinyl ether, 99%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3736509
18.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
109-92-2, 1310-58-3
C4H8O
72.11
MFCD00009248
FJKIXWOMBXYWOQ-UHFFFAOYSA-N
ethyl vinyl ether, ethoxyethene, ethoxyethylene, ethene, ethoxy, 1-ethoxyethene, 1-ethoxyethylene, vinyl ethyl ether, vinamar, ether, ethyl vinyl, ether, vinyl ethyl
8023
ethenoxyethane
CCOC=C
109-92-2, 1310-58-3
C4H8O
72.11
MFCD00009248
FJKIXWOMBXYWOQ-UHFFFAOYSA-N
ethyl vinyl ether, ethoxyethene, ethoxyethylene, ethene, ethoxy, 1-ethoxyethene, 1-ethoxyethylene, vinyl ethyl ether, vinamar, ether, ethyl vinyl, ether, vinyl ethyl
8023
ethenoxyethane
CCOC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -116.0°C
Color Colorless
Density 0.7500g/mL
Boiling Point 36.0°C
Flash Point −45°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OCH=CH2
Refractive Index 1.3760 to 1.3780
Quantity 250 mL
Beilstein 01,433
Fieser 01,386; 02,198; 04,234; 10,184; 11,235
Specific Gravity 0.75
Solubility Information Solubility in water: 7.8g/L (25°C). Other solubilities: soluble in most organic solvents
Stabilizer 0.1% KOH
Viscosity 0.28 mPa.s (5°C)
Formula Weight 72.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl vinyl ether, Stabilized
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'