Learn More
Saint Gobain Performance Plastics™ Ethyl Vinyl Acetate Microbore Tubing
Use this tubing for biological testing. Saint Gobain Performance Plastics™ Ethyl Vinyl Acetate Microbore Tubing is biologically stable, nontoxic and non-contaminating.
Brand:  Saint Gobain Performance Plastics™ 95712-30
Code : 50
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12428630
  • £55.63 / 1 roll
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Translucent cross-linked tubing flexible micro-line tubing
  • Virtually 100% elastic memory, returns to its original shape with gentle heating
  • Resin is USP 23 Class VI and National Formulary Vol. 14 specifications
  • Sterilize with autoclave, ethylene oxide, gamma irradiation or dry heat
Specifications
Specifications
Translucent Yellow
EVA
Solvent/Hydrocarbon Autoanalysis/Microbore color-coded stopped tubing
74°C
1 Pack
1.02mm
3.07mm
Solvent/Hydrocarbon Autoanalysis/Microbore color-coded stopped tubing
–50°C
30
Videos
Documentation