missing translation for 'onlineSavingsMsg'
Learn More

Ethyltriphenylphosphonium bromide, 98%, Acros Organics™

Brand:  Acros Organics 119041000

Code : 29

Additional Details : CAS Number : 1530-32-1 Weight : 0.10000kg Transport : UN number : 2811 Chem class : 1 Pack group : III


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10530591

  • £49.10 / 100g


EU Stock 5
Add to basket


Specifications

Chemical Identifiers

Ethyltriphenylphosphonium bromide
1530-32-1
100.0
98%
C2H5P(C6H5)3Br
MFCD00011838
99+%
97.5
98%
C20H20BrP
371.26

Specifications

Ethyltriphenylphosphonium bromide
White to off-white
100g
1530-32-1
100.0
Authentic
Crystalline Powder
98%
C2H5P(C6H5)3Br
MFCD00011838
>200°C
206.5° to 208.5°C
99+%
97.5
1% max.
Glass bottle
98%
C20H20BrP
371.26
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethyltriphenylphosphonium bromide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS