Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 265650050

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 28250-14-8 Weight: 0.05500kg


Packaging Glass bottle
Quantity 5g

Product Code. 11955341

Quantity Price
1 £ 380.8 / 5g
Estimated Shipment date
from Supplier 09-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride
Assay ≥97.5% (Argentometry)
Assay Percent Range 98%
Molecular Formula C9H17NO2·HCl
Formula Weight 207.7

Specification

Chemical Name or Material Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride
Color White to almost white
Melting Point 152° to 156°C
Quantity 5g
CAS 28250-14-8
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Granular Powder
Assay ≥97.5% (Argentometry)
Assay Percent Range 98%
Molecular Formula C9H17NO2·HCl
Formula Weight 207.7
Documentation

Technical documents