missing translation for 'onlineSavingsMsg'
Learn More

Ethyl tiglate, 98%, Thermo Scientific Chemicals

Catalog Number p-7081858
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
5837-78-5
C7H12O2
128.171
MFCD00015183
OAPHLAAOJMTMLY-GQCTYLIASA-N
ethyl tiglate, tiglic acid, ethyl ester, ethyl 2-methylcrotonate, ethyl alpha-methylcrotonate, ethyl trans-2-methyl-2-butenoate, e-2-methyl-2-butenoic acid ethyl ester, ethyl e-2-methylcrotonate, ethyl trans-2-methylcrotonate, tiglic acid ethyl ester, ethyl 2-methylbut-2-enoate
5281163
CHEBI:4892
ethyl (E)-2-methylbut-2-enoate
CCOC(=O)C(=CC)C
This item is not returnable. View return policy
5837-78-5
C7H12O2
128.171
MFCD00015183
OAPHLAAOJMTMLY-GQCTYLIASA-N
ethyl tiglate, tiglic acid, ethyl ester, ethyl 2-methylcrotonate, ethyl alpha-methylcrotonate, ethyl trans-2-methyl-2-butenoate, e-2-methyl-2-butenoic acid ethyl ester, ethyl e-2-methylcrotonate, ethyl trans-2-methylcrotonate, tiglic acid ethyl ester, ethyl 2-methylbut-2-enoate
5281163
CHEBI:4892
ethyl (E)-2-methylbut-2-enoate
CCOC(=O)C(=CC)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.93
Boiling Point 156°C
Flash Point 46°C (114°F)
Linear Formula CH3CH=C(CH3)CO2CH2CH3
Refractive Index 1.434
Quantity 25 g
UN Number UN3272
Beilstein 1720895
Merck Index 14,9433
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material Ethyl tiglate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'