missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl thiomorpholine-3-carboxylate, 97%

Brand:  Alfa Aesar™ H64122.03

Code : NEW

Additional Details : CAS Number : 58729-31-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15421179

  • £207.76 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl thiomorpholine-3-carboxylate
175.25
879254
58729-31-0
MFCD00107967
Thiomorpholine-3-carboxylic acid ethyl ester

Specifications

Ethyl thiomorpholine-3-carboxylate
1g
Solid
175.25
879254
118° to 121°C (8.5mmHg)
58729-31-0
97%
MFCD00107967
Thiomorpholine-3-carboxylic acid ethyl ester
Videos
SDS