missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl sorbate, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
The addition of chlorine and bromine to ethyl sorbate (la) gave 1, 2-and 1, a-dihalo products derived from attack of the halogen. Chlorination of la under ionic conditions proceeds through a tightly bridged chloronium ion intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The addition of chlorine and bromine to ethyl sorbate (la) gave 1, 2-and 1, a-dihalo products derived from attack of the halogen. Chlorination of la under ionic conditions proceeds through a tightly bridged chloronium ion intermediate.
Solubility
Soluble in alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
The addition of chlorine and bromine to ethyl sorbate (la) gave 1, 2-and 1, a-dihalo products derived from attack of the halogen. Chlorination of la under ionic conditions proceeds through a tightly bridged chloronium ion intermediate.
Solubility
Soluble in alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Density | 0.948 |
Boiling Point | 195°C to 196°C |
Flash Point | 69°C (156°F) |
Refractive Index | 1.495 |
Quantity | 25 g |
Beilstein | 1362941 |
Solubility Information | Soluble in alcohol. |
Formula Weight | 140.18 |
Percent Purity | 98% |
Chemical Name or Material | Ethyl sorbate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'