missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (R)-2-hydroxy-4-phenylbutyrate, 98%, ACROS Organics™

Brand:  Acros Organics 338870050

Code : 29

Additional Details : CAS Number : 90315-82-5 Weight : 0.06000kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10123151

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Ethyl (R)-2-hydroxy-4-phenylbutyrate
90315-82-5
98%
C6H5(CH2)2CH(OH)COOCH2CH3
MFCD00077794
98%
≥97.5%
C12H16O3
208.26

Specifications

Ethyl (R)-2-hydroxy-4-phenylbutyrate
90315-82-5
Authentic
Liquid
98%
1.5030 to 1.5050 (20°C, 589nm)
-9° to -11° (20°C, 589nm) (neat)
MFCD00077794
Clear colorless
98%
1.0750g/mL
Glass bottle
≥97.5%
C12H16O3
C6H5(CH2)2CH(OH)COOCH2CH3
208.26
− 10.00 (20.00°C c=neat)
5g
Videos
SDS