missing translation for 'onlineSavingsMsg'
Learn More

Ethyl pyrrolidinoacetate, 98%, ACROS Organics™

Brand:  Acros Organics 350920250

Code : 29

Additional Details : CAS Number : 22041-19-6 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10681453

  • £64.10 / 25g


Estimated Shipment
13-02-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl pyrrolidinoacetate
22041-19-6
98%
157.21
98%
≥97.5%
C8H15NO2
MFCD00012085

Specifications

Ethyl pyrrolidinoacetate
25g
22041-19-6
Glass bottle
Liquid
98%
157.21
20, V,1, 335
Clear light yellow
98%
Authentic
1.4460 to 1.4480 (20°C, 589nm)
≥97.5%
C8H15NO2
MFCD00012085
Videos
SDS