missing translation for 'onlineSavingsMsg'
Learn More

Ethyl propionate, Thermo Scientific Chemicals

Catalog Number p-7049593
Click to view available options
Quantity:
1 kg
3 x 1 kg
This item is not returnable. View return policy
105-37-3
C5H10O2
102.133
MFCD00009308
FKRCODPIKNYEAC-UHFFFAOYSA-N
ethyl propionate, propanoic acid, ethyl ester, propionic ester, propionic ether, propionic acid, ethyl ester, ethylpropionate, propionate d'ethyle, propanoic acid ethyl ester, ethyl n-propionate, propionic acid ethyl ester
7749
CHEBI:41330
ethyl propanoate
CCC(=O)OCC
This item is not returnable. View return policy
105-37-3
C5H10O2
102.133
MFCD00009308
FKRCODPIKNYEAC-UHFFFAOYSA-N
ethyl propionate, propanoic acid, ethyl ester, propionic ester, propionic ether, propionic acid, ethyl ester, ethylpropionate, propionate d'ethyle, propanoic acid ethyl ester, ethyl n-propionate, propionic acid ethyl ester
7749
CHEBI:41330
ethyl propanoate
CCC(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -73°C
Density 0.891 g/mL
Boiling Point 99°C
Flash Point 12°C (54°F)
Refractive Index 1.385
Quantity 1 kg
UN Number UN1195
Beilstein 506287
Merck Index 14,3847
Formula Weight 102.13
Physical Form Liquid
Chemical Name or Material Ethyl propionate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'