missing translation for 'onlineSavingsMsg'
Learn More

Ethyl propenyl ether, 98%, mixture of cis- and trans-isomers, Thermo Scientific Chemicals

Catalog Number p-2691472
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
928-55-2
C5H10O
86.13
MFCD00015184
XDHOEHJVXXTEDV-HYXAFXHYSA-N
1-ethoxypropene, ethyl propenyl ether, ether, ethyl propenyl, propenyl ethyl ether, e-1-ethoxyprop-1-ene, 1-ethoxy-1-propene, unii-jh817due12, ethyl 1-propenyl ether, 1-propene, 1-ethoxy, ethyl-1-propenyl ether
5365091
(E)-1-ethoxyprop-1-ene
CCO\C=C/C
This item is not returnable. View return policy
928-55-2
C5H10O
86.13
MFCD00015184
XDHOEHJVXXTEDV-HYXAFXHYSA-N
1-ethoxypropene, ethyl propenyl ether, ether, ethyl propenyl, propenyl ethyl ether, e-1-ethoxyprop-1-ene, 1-ethoxy-1-propene, unii-jh817due12, ethyl 1-propenyl ether, 1-propene, 1-ethoxy, ethyl-1-propenyl ether
5365091
(E)-1-ethoxyprop-1-ene
CCO\C=C/C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.7780g/mL
Boiling Point 67.0°C to 76.0°C
Flash Point −18°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CHCHOCH2CH3
Refractive Index 1.3940 to 1.4000
Quantity 10 g
Fieser 01,385
Specific Gravity 0.778
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alkanes,methylene chloride,ethanol,,2-propanol,acetone
Formula Weight 86.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl propenyl ether, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl propenyl ether, mixture of cis- and trans-isomers
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'