Ethyl Phenylpropiolate 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 118850250

25GR Ethyl phenylpropiolate, 98%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 2216-94-6 Weight: 0.07500kg


Packaging Glass bottle
Quantity 25g

Product Code. 10580751

Quantity Price
1 £ 55.6 / 25g
EU Stock 5
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethyl phenylpropiolate
Name Note 98%
CAS 2216-94-6
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H10O2
Linear Formula C6H5CCCO2C2H5
Formula Weight 174.2
MDL Number MFCD00009185

Specification

Physical Form Liquid
Assay ≥97.5%
Formula Weight 174.2
Quantity 25g
Color Clear yellow
Beilstein 09, 634
EINECS Number 218-703-8
Linear Formula C6H5CCCO2C2H5
CAS 2216-94-6
Infrared Spectrum Authentic
Refractive Index 1.5510 to 1.5530 (20°C, 589nm)
Assay Percent Range 98%
MDL Number MFCD00009185
RTECS Number UE0110000
Packaging Glass bottle
Name Note 98%
Molecular Formula C11H10O2
Density 1.05g/mL
Chemical Name or Material Ethyl phenylpropiolate
Documentation

Technical documents