missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenylpropiolate, 98%, Thermo Scientific Chemicals

Catalog Number p-131927
15.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2216-94-6
C11H10O2
174.2
MFCD00009185
ACJOYTKWHPEIHW-UHFFFAOYSA-N
ethyl phenylpropiolate, ethylphenylpropiolate, ethyl 3-phenylpropiolate, ethyl phenylpropriolate, 2-propynoic acid, 3-phenyl-, ethyl ester, ethyl 3-phenylpropynoate, ethyl phenylpropynoate, ethyl phenylacetylenecarboxylate, phenylpropiolic acid ethyl ester, ethyl 3-phenyl-2-propynoate
91516
ethyl 3-phenylprop-2-ynoate
CCOC(=O)C#CC1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2216-94-6
C11H10O2
174.2
MFCD00009185
ACJOYTKWHPEIHW-UHFFFAOYSA-N
ethyl phenylpropiolate, ethylphenylpropiolate, ethyl 3-phenylpropiolate, ethyl phenylpropriolate, 2-propynoic acid, 3-phenyl-, ethyl ester, ethyl 3-phenylpropynoate, ethyl phenylpropynoate, ethyl phenylacetylenecarboxylate, phenylpropiolic acid ethyl ester, ethyl 3-phenyl-2-propynoate
91516
ethyl 3-phenylprop-2-ynoate
CCOC(=O)C#CC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Density 1.0500g/mL
Boiling Point 260°C to 270°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5C∽CCO2C2H5
Refractive Index 1.5510 to 1.553
Quantity 5 g
Beilstein 09, 634
Specific Gravity 1.05
Formula Weight 174.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl phenylpropiolate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'