missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenylcyanoacetate, 97%, Thermo Scientific Chemicals

Catalog Number p-131918
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4553-07-5
C11H11NO2
189.21
MFCD00001866
SXIRJEDGTAKGKU-UHFFFAOYNA-N
CCOC(=O)C(C#N)C1=CC=CC=C1
This item is not returnable. View return policy
4553-07-5
C11H11NO2
189.21
MFCD00001866
SXIRJEDGTAKGKU-UHFFFAOYNA-N
CCOC(=O)C(C#N)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 275°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5CH(CN)CO2C2H5
Refractive Index 1.5033 to 1.5053
Quantity 5 g
Beilstein 09, 854
Specific Gravity 1.09
Synonym s-dibenzyl 2-aminopentanedioate hydrochloride, h-glu obzl-obzl.hcl, l-glutamic acid dibenzyl ester hydrochloride, l-glutamic acid, bis phenylmethyl ester, hydrochloride, s-dibenzyl 2-aminopentanedioate hcl, dibenzyl l-glutaminate hydrochloride, glutamic acid, dibenzyl ester, hydrochloride, l, 1,5-dibenzyl 2s-2-aminopentanedioate hydrochloride, h-glu obzl-obzl.hci, h-glu obzl-obzl hcl
IUPAC Name dibenzyl (2S)-2-aminopentanedioate;hydrochloride
PubChem CID 199613
Formula Weight 189.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl phenylcyanoacetate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl phenylcyanoacetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'