missing translation for 'onlineSavingsMsg'
Learn More

Ethyl p-toluate, 98+%, Thermo Scientific Chemicals

Catalog Number p-7080767
Click to view available options
Quantity:
50 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
94-08-6
C10H12O2
164.204
MFCD00009117
NWPWRAWAUYIELB-UHFFFAOYSA-N
ethyl p-toluate, ethyl p-methylbenzoate, benzoic acid, 4-methyl-, ethyl ester, 4-methylbenzoic acid ethyl ester, unii-xuf0sq8l2j, p-toluic acid ethyl ester, xuf0sq8l2j, p-toluic acid, ethyl ester, 4-methyl-benzoic acid ethyl ester, ethyl4-methylbenzoate
66743
ethyl 4-methylbenzoate
CCOC(=O)C1=CC=C(C=C1)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
94-08-6
C10H12O2
164.204
MFCD00009117
NWPWRAWAUYIELB-UHFFFAOYSA-N
ethyl p-toluate, ethyl p-methylbenzoate, benzoic acid, 4-methyl-, ethyl ester, 4-methylbenzoic acid ethyl ester, unii-xuf0sq8l2j, p-toluic acid ethyl ester, xuf0sq8l2j, p-toluic acid, ethyl ester, 4-methyl-benzoic acid ethyl ester, ethyl4-methylbenzoate
66743
ethyl 4-methylbenzoate
CCOC(=O)C1=CC=C(C=C1)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.025
Boiling Point 232°C
Flash Point 99°C (210°F)
Refractive Index 1.508
Quantity 50 g
Beilstein 1863756
Formula Weight 164.21
Percent Purity ≥98%
Chemical Name or Material Ethyl p-toluate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'