missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl oxazole-5-carboxylate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Ethyl oxazole-5-carboxylate is used as a pharmaceutical, organic intermediate and in organic light-emitting diode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl oxazole-5-carboxylate is used as a pharmaceutical, organic intermediate and in organic light-emitting diode.
Solubility
Soluble in water (35 g/L at 25°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Ethyl oxazole-5-carboxylate is used as a pharmaceutical, organic intermediate and in organic light-emitting diode.
Solubility
Soluble in water (35 g/L at 25°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Boiling Point | 60°C (2.25 mmHg) |
Refractive Index | 1.469 |
Quantity | 5 g |
Beilstein | 83084416 |
Solubility Information | Soluble in water (35g/L at 25°C). |
Formula Weight | 141.13 |
Percent Purity | 98% |
Chemical Name or Material | Ethyl oxazole-5-carboxylate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'