missing translation for 'onlineSavingsMsg'
Learn More

Ethyl orange sodium salt, 92%, ACROS Organics™

Brand:  Acros Organics 410300250

UNSPSC : 12000000

Code : 29

Additional Details : CAS Number : 62758-12-7 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10229940

  • £50.60 / 25g


Estimated Shipment
21-08-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

Ethyl Orange sodium salt
62758-12-7
92%
355.38
Pure
Pure
C16H18N3NaO3S
MFCD00007503

Specifications

Ethyl Orange sodium salt
62758-12-7
Crystalline Powder
C16H18N3NaO3S
MFCD00007503
Glass bottle
from pH 3.5 (red) to pH 4.8 (yellow)
25g
Pure
Pure
92%
355.38
Authentic
90% min.
Orange

Safety and Handling

Product Identifier
  • Ethyl Orange sodium salt

Documentation

Technical Documents