Alfa Aesar™ Ethyl O-[4-bromo-3-(trifluoromethyl)benzenesulfonyl]acetohydroxamate, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H63747.06

UNSPSC: 12352105

Code: NEW

Additional Details:
Additional Details: Weight: 0.00500kg


Quantity 5g

Product Code. 15438496

Quantity Price
1 £ 627.52 / 5g
Estimated Shipment date
from Supplier 13-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethyl O-[4-bromo-3-(trifluoromethyl)benzenesulfonyl]acetohydroxamate
Formula Weight 390.17
MDL Number MFCD27665234
Synonym O-[4-Bromo-3-(trifluoromethyl)benzenesulfonyl]acetohydroxamic acid ethyl ester

Specification

Formula Weight 390.17
Quantity 5g
Chemical Name or Material Ethyl O-[4-bromo-3-(trifluoromethyl)benzenesulfonyl]acetohydroxamate
Physical Form Solid
Assay 97%
MDL Number MFCD27665234
Synonym O-[4-Bromo-3-(trifluoromethyl)benzenesulfonyl]acetohydroxamic acid ethyl ester
Documentation

Technical documents