missing translation for 'onlineSavingsMsg'
Learn More

Ethyl nitroacetate, 98%, Thermo Scientific Chemicals

Catalog Number 10718352
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
626-35-7
C4H7NO4
133.1
FTKASJMIPSSXBP-UHFFFAOYSA-N
ethyl nitroacetate, nitroacetic acid ethyl ester, acetic acid, nitro-, ethyl ester, nitro ethyl acetate, ethylnitroacetate, acetic acid, 2-nitro-, ethyl ester, ethylnitroacetat, ethyinitroacetate, ethyl-2-nitroacetate, zlchem 269
69379
ethyl 2-nitroacetate
CCOC(=O)C[N+](=O)[O-]
This item is not returnable. View return policy
626-35-7
C4H7NO4
133.1
FTKASJMIPSSXBP-UHFFFAOYSA-N
ethyl nitroacetate, nitroacetic acid ethyl ester, acetic acid, nitro-, ethyl ester, nitro ethyl acetate, ethylnitroacetate, acetic acid, 2-nitro-, ethyl ester, ethylnitroacetat, ethyinitroacetate, ethyl-2-nitroacetate, zlchem 269
69379
ethyl 2-nitroacetate
CCOC(=O)C[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.1990g/mL
Boiling Point 105.0°C to 107.0°C (25.0 mmHg)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NCH2CO2C2H5
Refractive Index 1.4235 to 1.4255
Quantity 25 g
Beilstein 02,225
Specific Gravity 1.199
Formula Weight 133.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl nitroacetate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl nitroacetate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'