missing translation for 'onlineSavingsMsg'
Learn More

Ethyl nitroacetate, 98%, ACROS Organics™

Brand:  Acros Organics 185170050

Code : 29

Additional Details : CAS Number : 626-35-7 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10566821

  • £31.30 / 5g


EU Stock 4
Add to basket


Specifications

Chemical Identifiers

Ethyl nitroacetate
98%
O2NCH2CO2C2H5
98%
C4H7NO4
133.1

Specifications

Ethyl nitroacetate
105.0° to 107.0°C (25.0 mmHg)
5g
Authentic
Glass bottle
1.199
98%
C4H7NO4
133.1
92°C
Clear colorless to slightly yellow
626-35-7
1.1990g/mL
1.4235 to 1.4255
Liquid
98%
O2NCH2CO2C2H5
Videos
SDS