Learn More
Ethyl morpholine-2-carboxylate, 95%, Maybridge
Brand: Maybridge MO08391DA
Code : 31
Additional Details : CAS Number : 107904-06-3 Weight : 0.05100kg Transport : UN number : 2735 Chem class : 8 Pack group : III
Packaging | Amber Glass Bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
107904-06-3 | |
159.185 | |
PTWKMUDNOPBYJO-UHFFFAOYSA-N | |
15651197 | |
CCOC(=O)C1CNCCO1 |
C7H13NO3 | |
MFCD08751336 | |
ethyl 2-morpholinecarboxylate, 2-morpholinecarboxylic acid ethyl ester, 2-morpholinecarboxylicacid, ethyl ester, 2-morpholinecarboxylic acid, ethyl ester, morpholine-2-carboxylic acid ethyl ester, acmc-209xhl, 2-ethoxycarbonyl morpholine | |
ethyl morpholine-2-carboxylate |
Specifications
Ethyl morpholine-2-carboxylate | |
1g | |
95.0 | |
C7H13NO3 | |
ethyl 2-morpholinecarboxylate, 2-morpholinecarboxylic acid ethyl ester, 2-morpholinecarboxylicacid, ethyl ester, 2-morpholinecarboxylic acid, ethyl ester, morpholine-2-carboxylic acid ethyl ester, acmc-209xhl, 2-ethoxycarbonyl morpholine | |
CCOC(=O)C1CNCCO1 | |
159.185 | |
159.18 |
Amber Glass Bottle | |
107904-06-3 | |
95% | |
MFCD08751336 | |
PTWKMUDNOPBYJO-UHFFFAOYSA-N | |
ethyl morpholine-2-carboxylate | |
15651197 | |
95% |