missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Maybridge Ethyl morpholine-2-carboxylate, 10g, 95%

Brand:  Thermo Scientific Maybridge MO08391EA

Ethyl morpholine-2-carboxylate, 10g, 95%

Code : 31

Additional Details : CAS Number : 107904-06-3 Weight : 0.06000kg Transport : UN number : 2735 Chem class : 8 Pack group : III

Product Code. 10397724

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.