missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Methyl Ketone, Extra Pure, SLR, Fisher Chemical™

C4H8O, CAS Number-78-93-3, methyl ethyl ketone, methylethylketone, methyl acetone, 2-butanone, methylethyl ketone, meetco, butanone, 3-butanone, ethyl methyl ketone, ethylmethylketon, 2.5L, 80 deg.C, CHEBI:28398, Colorless, 72.11g/mol, ZWEHNKRNPOVVGH-UHFFFAOYSA-N, butan-2-one, -87 deg.C, 11648

Brand:  Fisher Chemical E/1450/PB17

59.03 GBP valid until 2021-02-28
Use promo code "14802" to get your promotional price.



Code : 21

Additional Details : CAS Number : 78-93-3 Weight : 2.01250kg Transport : UN number : 1193 Chem class : 3 Pack group : II


Packaging Plastic Coated Glass bottle
Quantity 2.5L

 View more versions of this product

Product Code. 10010130

  • £73.79 / 2.50 L

In stock, ready to ship: 25

Please to purchase this item. Need a web account? Register with us today!


Description

Description

C4H8O, CAS Number-78-93-3, methyl ethyl ketone, methylethylketone, methyl acetone, 2-butanone, methylethyl ketone, meetco, butanone, 3-butanone, ethyl methyl ketone, ethylmethylketon, 2.5L, 80 deg.C, CHEBI:28398, Colorless, 72.11g/mol, ZWEHNKRNPOVVGH-UHFFFAOYSA-N, butan-2-one, -87 deg.C, 11648

Specifications

Chemical Identifiers

78-93-3
72.107
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
6569
butan-2-one
C4H8O
11648
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
CHEBI:28398
CCC(=O)C

Specifications

Plastic Coated Glass bottle
Colorless
2.5L
C4H8O
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
CCC(=O)C
72.107
CHEBI:28398
Liquid
80°C
-87°C
78-93-3
11648
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
butan-2-one
6569
72.11g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Butanone

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P240-Ground/bond container and receiving equipment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents