missing translation for 'onlineSavingsMsg'
Learn More

Ethyl levulinate, 98%, Thermo Scientific Chemicals

Catalog Number p-130877
18.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
539-88-8
C7H12O3
144.17
MFCD00009209
GMEONFUTDYJSNV-UHFFFAOYSA-N
ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate
10883
ethyl 4-oxopentanoate
CCOC(=O)CCC(C)=O
This item is not returnable. View return policy
539-88-8
C7H12O3
144.17
MFCD00009209
GMEONFUTDYJSNV-UHFFFAOYSA-N
ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate
10883
ethyl 4-oxopentanoate
CCOC(=O)CCC(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 5mg KOH/g max.
Color Yellow
Density 1.0120g/mL
Boiling Point 206°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH2CH2CO2C2H5
Refractive Index 1.4210 to 1.423
Quantity 100 g
Beilstein 03, 675
Merck Index 15, 3874
Specific Gravity 1.012
Solubility Information Solubility in water: 40g/L(20°C). Other solubilities: miscible with alcohol
Formula Weight 144.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl levulinate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'