missing translation for 'onlineSavingsMsg'
Learn More

Ethyl L(-)-lactate, 97%, ACROS Organics™

£15.50 - £329.00

Chemical Identifiers

CAS 687-47-8
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00004518
InChI Key LZCLXQDLBQLTDK-BYPYZUCNSA-N
Synonym ethyl l-lactate, ethyl 2s-2-hydroxypropanoate, l-ethyl lactate, l---ethyl lactate, ethyl l--lactate, ethyl s-2-hydroxypropionate, ethyl l---lactate, s-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s
PubChem CID 92831
ChEBI CHEBI:78322
IUPAC Name ethyl (2S)-2-hydroxypropanoate
SMILES CCOC(=O)C(C)O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10561774
SDS View Documents
Acros Organics
118662500
250mL Glass bottle
£15.50
250mL
View alternative products
Save Up to 
»
In stock, ready to ship: 8
Please to purchase this item. Need a web account? Register with us today!
10142290
SDS View Documents
Acros Organics
118660010
1L Glass bottle
£42.20
1 L
View alternative products
Save Up to 
»
In stock, ready to ship: 17
Please to purchase this item. Need a web account? Register with us today!
10042353
SDS View Documents
Acros Organics
118660025
2.5L Glass bottle
£87.00
2.50 L
View alternative products
Save Up to 
»
In stock, ready to ship: 3
Please to purchase this item. Need a web account? Register with us today!
13418328
SDS View Documents
Acros Organics
118660100
10L Plastic drum
£329.00
10 L
View alternative products
Save Up to 
»
Estimated Shipment: 13-10-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

687-47-8
118.13
LZCLXQDLBQLTDK-BYPYZUCNSA-N
92831
ethyl (2S)-2-hydroxypropanoate
C5H10O3
MFCD00004518
ethyl l-lactate, ethyl 2s-2-hydroxypropanoate, l-ethyl lactate, l---ethyl lactate, ethyl l--lactate, ethyl s-2-hydroxypropionate, ethyl l---lactate, s-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s
CHEBI:78322
CCOC(=O)C(C)O

Specifications

0.1% max. (as lactic acid)
1.0340g/mL
Authentic
1.4100 to 1.4160
− 10.00 (20.00°C neat)
0.3% max. (K.F.)
Undesignated
250mL
96% min. (GC)
CH3CH(OH)CO2C2H5
03,264
14,3817
Solubility in water: soluble. Other solubilities: miscible with alcohols,ketones and esters
CCOC(=O)C(C)O
118.13
CHEBI:78322
Liquid
Ethyl L(-)-lactate
46°C
Glass bottle
1.034
− 10.00
154.0°C
-26.0°C
687-47-8
C5H10O3
MFCD00004518
17,135
ethyl l-lactate, ethyl 2s-2-hydroxypropanoate, l-ethyl lactate, l---ethyl lactate, ethyl l--lactate, ethyl s-2-hydroxypropionate, ethyl l---lactate, s-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s
LZCLXQDLBQLTDK-BYPYZUCNSA-N
ethyl (2S)-2-hydroxypropanoate
92831
118.13
97%
Safety and Handling

Safety and Handling

Product Identifier
  • Ethyl L(-)-lactate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Product Certifications