missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isovalerate, 99%, Thermo Scientific Chemicals

Catalog Number 10796371
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
108-64-5
C7H14O2
130.19
MFCD00009203
PPXUHEORWJQRHJ-UHFFFAOYSA-N
ethyl isovalerate, ethyl 3-methylbutyrate, ethylisovalerate, ethyl isopentanoate, isovaleric acid, ethyl ester, butanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylbutyrate, ethyl isovalerianate, 3-methylbutyric acid ethyl ester, 3-methylbutanoic acid ethyl ester
7945
CHEBI:31571
ethyl 3-methylbutanoate
CCOC(=O)CC(C)C
This item is not returnable. View return policy
108-64-5
C7H14O2
130.19
MFCD00009203
PPXUHEORWJQRHJ-UHFFFAOYSA-N
ethyl isovalerate, ethyl 3-methylbutyrate, ethylisovalerate, ethyl isopentanoate, isovaleric acid, ethyl ester, butanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylbutyrate, ethyl isovalerianate, 3-methylbutyric acid ethyl ester, 3-methylbutanoic acid ethyl ester
7945
CHEBI:31571
ethyl 3-methylbutanoate
CCOC(=O)CC(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -99°C
Color Colorless to Yellow
Density 0.8630g/mL
Boiling Point 131°C to 133°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CO2C2H5
Refractive Index 1.3950 to 1.398
Quantity 100 mL
Beilstein 02, 312
Merck Index 15, 3871
Specific Gravity 0.863
Solubility Information Solubility in water: < 1%. Other solubilities: miscible with alcohol,benzene and ether
Formula Weight 130.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl isovalerate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl isovalerate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'