missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isodehydroacetate, 95%, Thermo Scientific™

Catalog Number p-4813477
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3385-34-0
C10H12O4
196.20
MFCD00006643
FBPWNVQUVXSXKS-UHFFFAOYSA-N
ethyl isodehydroacetate, ethyl 4,6-dimethyl-2-oxo-2h-pyran-5-carboxylate, ethyl isodehydracetate, 5-carbethoxy-4,6-dimethyl-2-pyrone, 2h-pyran-5-carboxylic acid, 4,6-dimethyl-2-oxo-, ethyl ester, ethyl 4,6-dimethyl-5-coumalate, 2,4-dimethyl-6-oxo-6h-pyran-3-carboxylic acid ethyl ester, ethylisodehydroacetate, ethyl 4,6-dimethyl-2-oxopyran-5-carboxylate, acmc-20am57
76918
ethyl 2,4-dimethyl-6-oxopyran-3-carboxylate
CCOC(=O)C1=C(C)OC(=O)C=C1C
This item is not returnable. View return policy
3385-34-0
C10H12O4
196.20
MFCD00006643
FBPWNVQUVXSXKS-UHFFFAOYSA-N
ethyl isodehydroacetate, ethyl 4,6-dimethyl-2-oxo-2h-pyran-5-carboxylate, ethyl isodehydracetate, 5-carbethoxy-4,6-dimethyl-2-pyrone, 2h-pyran-5-carboxylic acid, 4,6-dimethyl-2-oxo-, ethyl ester, ethyl 4,6-dimethyl-5-coumalate, 2,4-dimethyl-6-oxo-6h-pyran-3-carboxylic acid ethyl ester, ethylisodehydroacetate, ethyl 4,6-dimethyl-2-oxopyran-5-carboxylate, acmc-20am57
76918
ethyl 2,4-dimethyl-6-oxopyran-3-carboxylate
CCOC(=O)C1=C(C)OC(=O)C=C1C

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 17°C to 18°C
Density 1.1670g/mL
Boiling Point 290°C to 295°C
Flash Point >110°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5135 to 1.5165
Quantity 5 g
Beilstein 18,41
Specific Gravity 1.167
Solubility Information Solubility in water: insoluble
Formula Weight 196.2
Percent Purity 95%
Chemical Name or Material Ethyl isodehydroacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'