missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isocyanatoacetate, 98%, Thermo Scientific Chemicals

Catalog Number p-4089662
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2949-22-6
C5H7NO3
129.12
MFCD00002040
DUVOZUPPHBRJJO-UHFFFAOYSA-N
ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester
76283
ethyl 2-isocyanatoacetate
CCOC(=O)CN=C=O
This item is not returnable. View return policy
2949-22-6
C5H7NO3
129.12
MFCD00002040
DUVOZUPPHBRJJO-UHFFFAOYSA-N
ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester
76283
ethyl 2-isocyanatoacetate
CCOC(=O)CN=C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.1510g/mL
Boiling Point 67°C to 68°C (11.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula OCNCH2COOC2H5
Refractive Index 1.4210 to 1.4230 (20°C, 589nm)
Quantity 5 g
Specific Gravity 1.151
Solubility Information Solubility in water: hydrolyses
Formula Weight 129.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl isocyanatoacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'