missing translation for 'onlineSavingsMsg'
Learn More

Ethyl indole-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog Number p-130314
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3770-50-1
C11H11NO2
189.21
MFCD00005609
QQXQAEWRSVZPJM-UHFFFAOYSA-N
ethyl indole-2-carboxylate, indole-2-carboxylic acid ethyl ester, 2-carbethoxyindole, 1h-indole-2-carboxylic acid, ethyl ester, indole-2-carboxylic acid, ethyl ester, 1h-indole-2-carboxylic acid ethyl ester, ethylindole-2-carboxylate, ethyl 2-indolecarboxylate, 1h-indole-2-carboxylic acid,ethyl ester, 2-carboethoxyindole
73125
ethyl 1H-indole-2-carboxylate
CCOC(=O)C1=CC2=CC=CC=C2N1
This item is not returnable. View return policy
3770-50-1
C11H11NO2
189.21
MFCD00005609
QQXQAEWRSVZPJM-UHFFFAOYSA-N
ethyl indole-2-carboxylate, indole-2-carboxylic acid ethyl ester, 2-carbethoxyindole, 1h-indole-2-carboxylic acid, ethyl ester, indole-2-carboxylic acid, ethyl ester, 1h-indole-2-carboxylic acid ethyl ester, ethylindole-2-carboxylate, ethyl 2-indolecarboxylate, 1h-indole-2-carboxylic acid,ethyl ester, 2-carboethoxyindole
73125
ethyl 1H-indole-2-carboxylate
CCOC(=O)C1=CC2=CC=CC=C2N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 119°C to 125°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 22, I, 506
Solubility Information Solubility in water: insoluble in water
Formula Weight 189.21
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Ethyl indole-2-carboxylate, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'