missing translation for 'onlineSavingsMsg'
Learn More

Ethyl heptafluorobutyrate, 97%, ACROS Organics™

Brand:  Acros Organics 174440250

Code : 29

Additional Details : CAS Number : 356-27-4 Weight : 0.07500kg Transport : UN number : 3272 Chem class : 1 Pack group : II


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10722031

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Ethyl heptafluorobutyrate
356-27-4
97%
CF3CF2CF2CO2C2H5
MFCD00000434
97%
≥96.0%
C6H5F7O2
242.09

Specifications

Ethyl heptafluorobutyrate
356-27-4
Glass bottle
≥96.0%
C6H5F7O2
CF3CF2CF2CO2C2H5
MFCD00000434
25g
97%
Authentic
Liquid
97%
1.3010 to 1.3030 (20°C, 589nm)
242.09
Clear colorless
Videos
SDS