missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl fluoroacetate, 98%, Thermo Scientific Chemicals
15.53 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Description
Ethyl fluoroacetate is used in the synthesis of 4-deoxy-4-fluoro-muscarines. It is also used as starting reagent for the synthesis of ethyl(diethoxyphosphoryl)fluoroacetate. and in development of an ammonolysis-based microencapsulation process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl fluoroacetate is used in the synthesis of 4-deoxy-4-fluoro-muscarines. It is also used as starting reagent for the synthesis of ethyl(diethoxyphosphoryl)fluoroacetate. and in development of an ammonolysis-based microencapsulation process.
Solubility
Decomposes in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Ethyl fluoroacetate is used in the synthesis of 4-deoxy-4-fluoro-muscarines. It is also used as starting reagent for the synthesis of ethyl(diethoxyphosphoryl)fluoroacetate. and in development of an ammonolysis-based microencapsulation process.
Solubility
Decomposes in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Color | Colorless to Yellow |
Density | 1.093 g/mL |
Boiling Point | 119°C to 120°C |
Linear Formula | FCH2CO2CH2CH3 |
Refractive Index | 1.375 |
Quantity | 10 g |
UN Number | UN2929 |
Beilstein | 1743761 |
Merck Index | 14,4167 |
Solubility Information | Decomposes in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'