missing translation for 'onlineSavingsMsg'
Learn More

Ethyl difluoroacetate, 97%, Thermo Scientific Chemicals

Catalog Number 10492673
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
454-31-9
C4H6F2O2
124.09
MFCD00013578
GZKHDVAKKLTJPO-UHFFFAOYSA-N
ethyl difluoroacetate, acetic acid, difluoro-, ethyl ester, difluoroacetic acid ethyl ester, ethyldifluoroacetate, ethyl difluoroacetic, ethyl difluoro acetate, difluoro-acetic acid ethyl ester, ethyl difluoroacetic acid, acetic acid, 2,2-difluoro-, ethyl ester, ethyldifluoracetat
9961
ethyl 2,2-difluoroacetate
CCOC(=O)C(F)F
This item is not returnable. View return policy
454-31-9
C4H6F2O2
124.09
MFCD00013578
GZKHDVAKKLTJPO-UHFFFAOYSA-N
ethyl difluoroacetate, acetic acid, difluoro-, ethyl ester, difluoroacetic acid ethyl ester, ethyldifluoroacetate, ethyl difluoroacetic, ethyl difluoro acetate, difluoro-acetic acid ethyl ester, ethyl difluoroacetic acid, acetic acid, 2,2-difluoro-, ethyl ester, ethyldifluoracetat
9961
ethyl 2,2-difluoroacetate
CCOC(=O)C(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.2420g/mL
Boiling Point 97.0°C
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula F2CHCO2CH2CH3
Refractive Index 1.3465 to 1.3485
Quantity 25 g
UN Number 2924
Specific Gravity 1.242
Formula Weight 124.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl difluoroacetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl difluoroacetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'