missing translation for 'onlineSavingsMsg'
Learn More

Ethyl diethoxyacetate, 98%, Thermo Scientific Chemicals

Catalog Number p-3559318
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
6065-82-3
C8H16O4
176.21
MFCD00009231
XCLBIKIQSCTANZ-UHFFFAOYSA-N
ethyl diethoxyacetate, acetic acid, diethoxy-, ethyl ester, diethoxyacetic acid ethyl ester, ethyldiethoxyacetate, acetic acid, 2,2-diethoxy-, ethyl ester, acetic acid,diethoxy,ethyl ester, acmc-209mlb, ethyl bis ethyloxy acetate, ethyl glyoxylate diethyl acetal
80169
ethyl 2,2-diethoxyacetate
CCOC(C(=O)OCC)OCC
This item is not returnable. View return policy
6065-82-3
C8H16O4
176.21
MFCD00009231
XCLBIKIQSCTANZ-UHFFFAOYSA-N
ethyl diethoxyacetate, acetic acid, diethoxy-, ethyl ester, diethoxyacetic acid ethyl ester, ethyldiethoxyacetate, acetic acid, 2,2-diethoxy-, ethyl ester, acetic acid,diethoxy,ethyl ester, acmc-209mlb, ethyl bis ethyloxy acetate, ethyl glyoxylate diethyl acetal
80169
ethyl 2,2-diethoxyacetate
CCOC(C(=O)OCC)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.9890g/mL
Boiling Point 198°C to 200°C (761.0 mmHg)
Flash Point 85°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula (C2H5O)2CHCO2C2H5
Refractive Index 1.41
Quantity 25 g
Beilstein 03, 601
Fieser 06,253
Specific Gravity 0.989
Formula Weight 176.21
Percent Purity 98%
Chemical Name or Material Ethyl diethoxyacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'