missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl diethoxyacetate, 97+%, Thermo Scientific Chemicals
26.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Ethyl diethoxyacetate is used in the preparation of a fatty acid analog containing DTS, 12,13-dioxotetradecanoic acid di(N-methyl-thiosemicarbazone) (FA-DTS). It is used for synthesis of heterocyclic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl diethoxyacetate is used in the preparation of a fatty acid analog containing DTS, 12,13-dioxotetradecanoic acid di(N-methyl-thiosemicarbazone) (FA-DTS). It is used for synthesis of heterocyclic compounds.
Solubility
Soluble in Dichloromethane, Ethyl Acetate and Methanol.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Ethyl diethoxyacetate is used in the preparation of a fatty acid analog containing DTS, 12,13-dioxotetradecanoic acid di(N-methyl-thiosemicarbazone) (FA-DTS). It is used for synthesis of heterocyclic compounds.
Solubility
Soluble in Dichloromethane, Ethyl Acetate and Methanol.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Density | 0.988 |
Boiling Point | 198°C to 200°C |
Flash Point | 85°C (185°F) |
Refractive Index | 1.41 |
Quantity | 25 g |
Beilstein | 878579 |
Solubility Information | Soluble in Dichloromethane,Ethyl Acetate and Methanol. |
Formula Weight | 176.21 |
Percent Purity | ≥97% |
Chemical Name or Material | Ethyl diethoxyacetate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'