missing translation for 'onlineSavingsMsg'
Learn More

Ethyl dichlorophosphate, 97%, Thermo Scientific Chemicals

Catalog Number p-128926
19.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1498-51-7
C2H5Cl2O2P
162.94
MFCD00002069
YZBOZNXACBQJHI-UHFFFAOYSA-N
ethyl dichlorophosphate, ethyl phosphorodichloridate, phosphorodichloridic acid, ethyl ester, ethyldichlorophosphate, dichloroethoxyphosphine oxide, ethylphosphoric acid dichloride, dichlorophosphoric acid, ethyl ester, unii-4ip5dtw75u, ethylesterdichlorid german, hsdb 417
15158
1-dichlorophosphoryloxyethane
CCOP(=O)(Cl)Cl
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1498-51-7
C2H5Cl2O2P
162.94
MFCD00002069
YZBOZNXACBQJHI-UHFFFAOYSA-N
ethyl dichlorophosphate, ethyl phosphorodichloridate, phosphorodichloridic acid, ethyl ester, ethyldichlorophosphate, dichloroethoxyphosphine oxide, ethylphosphoric acid dichloride, dichlorophosphoric acid, ethyl ester, unii-4ip5dtw75u, ethylesterdichlorid german, hsdb 417
15158
1-dichlorophosphoryloxyethane
CCOP(=O)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Brown
Density 1.3730g/mL
Boiling Point 167°C (758.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C2H5OP(O)Cl2
Refractive Index 1.4330 to 1.435
Quantity 5 g
Beilstein 01,332
Specific Gravity 1.373
Formula Weight 162.94
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl dichlorophosphate, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl dichlorophosphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'