missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl cyanoacetate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2500 g
Description
Cyanoacetate is a building block for the synthesis of heterocycles and are are used as drugs. It is also used as Pharmaceutical intermediates, or Oil soluble couplers of film raw material . It is also used as a reagent in labelled pyrimidine and purine synthesis. It is used as an intermediate of fine chemicals such as medicine and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyanoacetate is a building block for the synthesis of heterocycles and are are used as drugs. It is also used as Pharmaceutical intermediates, or Oil soluble couplers of film raw material . It is also used as a reagent in labelled pyrimidine and purine synthesis. It is used as an intermediate of fine chemicals such as medicine and dyestuff.
Solubility
It is slightly soluble in water. Mostly soluble in organic solvents such as chloroform, ethyl acetate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials strong acids, strong bases, strong oxidizing agents, strong reducing agents.
Cyanoacetate is a building block for the synthesis of heterocycles and are are used as drugs. It is also used as Pharmaceutical intermediates, or Oil soluble couplers of film raw material . It is also used as a reagent in labelled pyrimidine and purine synthesis. It is used as an intermediate of fine chemicals such as medicine and dyestuff.
Solubility
It is slightly soluble in water. Mostly soluble in organic solvents such as chloroform, ethyl acetate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials strong acids, strong bases, strong oxidizing agents, strong reducing agents.
Specifications
Melting Point | -23°C |
Density | 1.063 |
Boiling Point | 208°C to 210°C |
Flash Point | 109°C (228°F) |
Odor | Weak |
Linear Formula | NCCH2CO2CH2CH3 |
Refractive Index | 1.418 |
Quantity | 500 g |
Beilstein | 605871 |
Merck Index | 14,3786 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'