missing translation for 'onlineSavingsMsg'
Learn More

Ethyl chrysanthemate, 95%, Thermo Scientific™

Catalog Number 15465958
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
97-41-6
C12H20O2
196.29
MFCD00001304
VIMXTGUGWLAOFZ-UHFFFAOYSA-N
ethyl chrysanthemate, ethyl chrysanthemumate, chrysanthemic acid ethyl ester, cyclopropanecarboxylic acid, 2,2-dimethyl-3-2-methyl-1-propenyl-, ethyl ester, ethylchrysanthemate, ccris 2498, chrysanthemic acid, ethyl ester, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methylprop-1-en-1-yl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropane-1-carboxylate
7334
ethyl 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
CCOC(=O)C1C(C1(C)C)C=C(C)C
This item is not returnable. View return policy
97-41-6
C12H20O2
196.29
MFCD00001304
VIMXTGUGWLAOFZ-UHFFFAOYSA-N
ethyl chrysanthemate, ethyl chrysanthemumate, chrysanthemic acid ethyl ester, cyclopropanecarboxylic acid, 2,2-dimethyl-3-2-methyl-1-propenyl-, ethyl ester, ethylchrysanthemate, ccris 2498, chrysanthemic acid, ethyl ester, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methylprop-1-en-1-yl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropane-1-carboxylate
7334
ethyl 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
CCOC(=O)C1C(C1(C)C)C=C(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Yellow
Density 0.906 g/mL
Boiling Point 112°C (10 mmHg)
Flash Point 84°C (183°F)
Refractive Index 1.4615
Quantity 25 g
Beilstein 2045740
Formula Weight 196.29
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl chrysanthemate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'