missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl bromoacetate, 98%, Thermo Scientific Chemicals
16.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -38.0°C |
Density | 1.5060g/mL |
Boiling Point | 159.0°C |
Flash Point | 47°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C4H7BrO2 |
Linear Formula | BrCH2CO2C2H5 |
Refractive Index | 1.4500 to 1.4520 |
Show More |
Safety and Handling
Product Identifier
- Ethyl bromoacetate
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- Flammable liquid Category 3
- H310-Fatal in contact with skin.
- H300-Fatal if swallowed.
- H330-Fatal if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'