missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Benzoate, 99+%, Thermo Scientific Chemicals

Catalog Number p-127401
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
93-89-0
C9H10O2
150.18
MFCD00009109
MTZQAGJQAFMTAQ-UHFFFAOYSA-N
benzoic acid, ethyl ester, benzoic ether, ethyl benzenecarboxylate, ethylbenzoate, benzoyl ethyl ether, benzoic acid ethyl ester, ethylester kyseliny benzoove, natural, unii-j115brj15h, fema no. 2422
7165
ethyl benzoate
CCOC(=O)C1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
93-89-0
C9H10O2
150.18
MFCD00009109
MTZQAGJQAFMTAQ-UHFFFAOYSA-N
benzoic acid, ethyl ester, benzoic ether, ethyl benzenecarboxylate, ethylbenzoate, benzoyl ethyl ether, benzoic acid ethyl ester, ethylester kyseliny benzoove, natural, unii-j115brj15h, fema no. 2422
7165
ethyl benzoate
CCOC(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 1mg KOH/g max.
Melting Point -34.0°C
Color Colorless to Yellow
Density 1.0450g/mL
Boiling Point 212.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CO2C2H5
Refractive Index 1.5040 to 1.5060
Quantity 500 g
Beilstein 09, IV, 285
Merck Index 15, 3821
Specific Gravity 1.045
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and chloroform
Formula Weight 150.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethyl benzoate, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl benzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'