missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Alcohol, Reagent, Denat., Spectro Grade, Contains 5% 2-PrOH and 5% MeOH, ACROS Organics™

Brand:  Acros Organics 224090010

UNSPSC : 12191601

Code : 29

Additional Details : CAS Number : 64-17-5 Weight : 0.83000kg Transport : UN number : 1987 Chem class : 1 Pack group : II

Quantity 1L
Packaging Glass bottle

 View more versions of this product

Product Code. 10481071

  • £58.00 / 1 L


EU Stock 63
Add to basket


Specifications Description & Specifications

Chemical Identifiers

Ethanol
For spectroscopy
C2H6O
46.06
67-63-0
4.0 to 6.0% (2-propanol)
C2H5OH

Specifications

Liquid
1L
4.0 to 6.0% (2-propanol)
0.0005% max.
67-56-1
0.01 max. at 300nm
0.1% Max. (Coulometric)
1.59
14, 3761
46.06
Clear
Vapor Pressure: 59mbar at 20°C
C2H5OH
For spectroscopy
Glass bottle
C2H6O
Ethanol
Documentation

Technical Documents