missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -71°C |
Density | 0.9200g/mL |
Boiling Point | 99°C |
Flash Point | 16°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99.40% min. (GC) |
Packaging | Glass bottle |
Linear Formula | H2C=CHCO2C2H5 |
Refractive Index | 1.4050 to 1.407 |
Quantity | 25 mL |
Show More |
Safety and Handling
Product Identifier
- Ethyl acrylate, stabilized
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Acute toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H331-Toxic if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'