missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetoacetate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
2.5 L
Specifications
Melting Point | -45°C |
Color | Colorless |
pH | 4 |
Boiling Point | 180°C |
Packaging | Amber glass bottle |
Quantity | 2.5 L |
Vapor Density | 4.5 |
Formula Weight | 130.14g/mol |
Vapor Pressure | 1 hPa at 20°C |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'